
Heptane-1,7-diamine, N-BOC protected 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:99733-18-3
:
Formula:C₁₂H₂₆N₂O₂
MDL Number:MFCD02094498
:230.347 g/mol
Boiling Pt:96…100 °C (0.1 mm Hg)
Melting Pt:113…114 °C
:GHS07
Storage Temperature:Refrigerator