Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,7-Dibromo-1H-indazol-4-amine
5,7-Dibromo-1H-indazol-4-amine
5,7-Dibromo-1H-indazol-4-amine
:
Apollo Scientific
5,7-Dibromo-1H-indazol-4-amine
:
Apollo Scientific
:
1427460-73-8
:
:
C₇H₅Br₂N₃
:
MFCD23159162
:
290.943 g/mol
:
GHS07
:
Ambient