Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-4,5-dimethylthiophene-3-carboxylic acid
2-Amino-4,5-dimethylthiophene-3-carboxylic acid
2-Amino-4,5-dimethylthiophene-3-carboxylic acid
Irritant
2-Amino-4,5-dimethylthiophene-3-carboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Amino-4,5-dimethylthiophene-3-carboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
55502-96-0
:
Formula:
C₇H₉NO₂S
MDL Number:
MFCD00456549
:
171.217 g/mol
:
GHS07
Storage Temperature:
Refrigerator