Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(1,3-Benzodioxol-5-yl)-2-chloroacetamide
2-(1,3-Benzodioxol-5-yl)-2-chloroacetamide
2-(1,3-Benzodioxol-5-yl)-2-chloroacetamide
Irritant
2-(1,3-Benzodioxol-5-yl)-2-chloroacetamide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(1,3-Benzodioxol-5-yl)-2-chloroacetamide
Supplier:
Apollo Scientific
About this item
CAS Number:
873380-46-2
:
Formula:
C₉H₈ClNO₃
MDL Number:
MFCD08445595
:
213.618 g/mol
:
GHS07
Storage Temperature:
Ambient