2,4-Dichlorophenoxyacetamidoxime
Bottle, 250 mg
Catalog # APOSOR0358-250MG
Supplier: Apollo Scientific
2,4-Dichlorophenoxyacetamidoxime
Catalog # APOSOR0358-250MG
Supplier: Apollo Scientific
CAS Number: 79295-15-1
Jump to:
Product Details & Documents
Synonyms: 2-(2,4-Dichlorophenoxy)-N'-hydroxyacetamidine
- CAS Number:79295-15-1
- Formula:C₈H₈Cl₂N₂O₂
- MDL Number:MFCD02090614
- Molecular Weight:235.067 g/mol
- Melting Pt:141…144 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



