Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,4,5-Tetrahydro-1H-1,5-benzodiazepine
2,3,4,5-Tetrahydro-1H-1,5-benzodiazepine
2,3,4,5-Tetrahydro-1H-1,5-benzodiazepine
Irritant
2,3,4,5-Tetrahydro-1H-1,5-benzodiazepine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3,4,5-Tetrahydro-1H-1,5-benzodiazepine
Supplier:
Apollo Scientific
About this item
CAS Number:
6516-89-8
:
Formula:
C₉H₁₂N₂
MDL Number:
MFCD06245443
:
148.205 g/mol
:
GHS07
Storage Temperature:
Ambient