
N-(1-Benzylpiperidin-4-yl)benzene-1,2-diamine, tech
Catalog # APOSOR01657-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:281.395 g/mol
Melting Pt:111…112 °C
Storage Temperature:Ambient
EINECS:260-916-3
UN:3259
ADR:8,III
:GHS05
CAS Number:57718-47-5
Formula:C₁₈H₂₃N₃
MDL Number:MFCD00023740