2,3-Diphenylpiperazine
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:143699-24-5
- Formula:C₁₆H₁₈N₂
- MDL Number:MFCD06245414
- Molecular Weight:238.328 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
- CAS Number:143699-24-5
- Formula:C₁₆H₁₈N₂
- MDL Number:MFCD06245414
- Molecular Weight:238.328 g/mol
- Storage Temperature:Ambient



