Order Entry
Export
ContactUsLinkComponent
2,3-Dihydro-4-(3-hydroxyphenyl)-1H-1,5-benzodiazepine
2,3-Dihydro-4-(3-hydroxyphenyl)-1H-1,5-benzodiazepine
Catalog # APOSOR01589-50MG
Supplier:  Apollo Scientific
CAS Number:  
2,3-Dihydro-4-(3-hydroxyphenyl)-1H-1,5-benzodiazepine
Catalog # APOSOR01589-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :238.284 g/mol
Storage Temperature:Ambient
CAS Number:889955-12-8
Formula:C₁₅H₁₄N₂O
MDL Number:MFCD06245402