Order Entry
Export
ContactUsLinkComponent
2,3-Dihydro-4-(fur-2-yl)-1H-1,5-benzodiazepine
Irritant
2,3-Dihydro-4-(fur-2-yl)-1H-1,5-benzodiazepine
Catalog # APOSOR01519-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Dihydro-4-(fur-2-yl)-1H-1,5-benzodiazepine
Catalog # APOSOR01519-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :212.247 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:394655-12-0
Formula:C₁₃H₁₂N₂O
MDL Number:MFCD06245352