Order Entry
Export
ContactUsLinkComponent
25-Hydroxyvitamin-D3-[2H3]
25-Hydroxyvitamin-D3-[2H3]
Catalog # APOSBISC1029-1MG
Supplier:  Apollo Scientific
CAS Number:  
25-Hydroxyvitamin-D3-[2H3]
Catalog # APOSBISC1029-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :403.656 g/mol
Storage Temperature:Ambient
Formula:C₂₇H₄₁D₃O₂
MDL Number:MFCD11656128