0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Hydroxy-PEG-5-t-butyl ester
Hydroxy-PEG-5-t-butyl ester
Hydroxy-PEG-5-t-butyl ester
Irritant
Hydroxy-PEG-5-t-butyl ester
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Hydroxy-PEG-5-t-butyl ester
Supplier:
Apollo Scientific
About this item
CAS Number:
850090-09-4
:
Formula:
C₁₇H₃₄O₈
MDL Number:
MFCD20926402
:
366.447 g/mol
Density:
1.071 g/cm³ (25 °C)
:
GHS07
Storage Temperature:
Ambient