Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydroxy-PEG6-acid
Hydroxy-PEG6-acid
Hydroxy-PEG6-acid
Hydroxy-PEG6-acid
Supplier:
Apollo Scientific
Certificates
Expand image
Hydroxy-PEG6-acid
Supplier:
Apollo Scientific
About this item
CAS Number:
1347750-85-9
:
Formula:
C₁₅H₃₀O₉
MDL Number:
MFCD22574804
:
354.393 g/mol
Storage Temperature:
Ambient