Order Entry
Export
ContactUsLinkComponent
Biotin-PEG6-Amine
Irritant
Biotin-PEG6-Amine
Catalog # APOSBIPG1211-50MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Biotin-PEG6-Amine
Catalog # APOSBIPG1211-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :550.709 g/mol
 :GHS07
Storage Temperature:Refrigerator
Formula:C₂₄H₄₆N₄O₈S
MDL Number:MFCD27977503