Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Azido-PEG6-amine
Azido-PEG6-amine
Azido-PEG6-amine
Azido-PEG6-amine
APOSBIPG1129-1G
:
Apollo Scientific
:
Azido-PEG6-amine
APOSBIPG1129-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
350.411 g/mol
:
Refrigerator
:
957486-82-7
:
C₁₄H₃₀N₄O₆
:
MFCD03453240