Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amino-PEG6-alcohol
Amino-PEG6-alcohol
Amino-PEG6-alcohol
:
Apollo Scientific
Amino-PEG6-alcohol
:
Apollo Scientific
:
39160-70-8
:
:
C₁₂H₂₇NO₆
:
MFCD22200781
:
281.346 g/mol
:
GHS07
:
Ambient