Ald-Ph-PEG5-t-butyl ester
Catalog # APOSBIPG1026-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:426.501 g/mol
Storage Temperature:Ambient
CAS Number:1433996-83-8
Formula:C₂₂H₃₄O₈
MDL Number:MFCD24539463