Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
24-Epibrassinolide
24-Epibrassinolide
24-Epibrassinolide
Irritant
24-Epibrassinolide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
24-Epibrassinolide
Supplier:
Apollo Scientific
About this item
CAS Number:
78821-43-9
:
Formula:
C₂₈H₄₈O₆
MDL Number:
MFCD00133310
Melting Pt:
254…256 °C
:
480.677 g/mol
:
GHS07
Storage Temperature:
Freezer