Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-Anhydro-β-D-xylofuranose
1,5-Anhydro-β-D-xylofuranose ≥97%
1,5-Anhydro-β-D-xylofuranose ≥97%
Irritant
1,5-Anhydro-β-D-xylofuranose ≥97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,5-Anhydro-β-D-xylofuranose ≥97%
Supplier:
Apollo Scientific
About this item
CAS Number:
51246-91-4
:
Formula:
C₅H₈O₄
MDL Number:
MFCD28122838
:
132.115 g/mol
:
GHS07
Storage Temperature:
Ambient