
DTSSP (3,3'-Dithiobis(sulphosuccinimidyl propionate))
Catalog # APOSBICL103-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:564.542 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:81069-02-5
Formula:C₁₄H₁₆N₂O₁₄S₄
MDL Number:MFCD01861949