
Sulphosuccinimidyl-2-(biotinamido)ethyl-1,3-dithiopropionate
Catalog # APOSBIB105-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:656.77 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:858128-64-0
Formula:C₂₂H₃₂N₄O₁₁S₄
MDL Number:MFCD01861945