
Alamethicin (from Trichoderma viride)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:27061-78-5
Formula:C₉₂H₁₅₀N₂₂O₂₅
MDL Number:MFCD00151517
:1964.40 g/mol
Melting Pt:272…278 °C
Storage Temperature:Freezer
UN:3462
ADR:6.1,III
:GHS06