0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Fluorinert™
Fluorinert™ FC-70
Fluorinert™ FC-70
:
Apollo Scientific
Fluorinert™ FC-70
:
Apollo Scientific
:
86508-42-1
:
311-89-7
:
MFCD09753882
:
201…221 °C
:
–25 °C
:
1.94 g/cm3 @ 25°C
:
GHS07
:
Ambient