Polyethylene glycol 300, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
:300 g/mol
Specification Test Results
- Hydroxyl value:356 - 394
- pH (5%):5,0 - 7,0
- Viscosity (20°C):80,0-105,0 mPa.s
- Heavy metals (as Pb):max 5 ppm
- Ignition residue (SO4):max 0.1 %
- Water:max 0.5%