Order Entry
Export
ContactUsLinkComponent
Platinum nominally 60% on carbon black
Platinum nominally 60% on carbon black
Catalog # 44171.18
CAS Number:  
Platinum nominally 60% on carbon black
Catalog # 44171.18
CAS Number:  

Specifications

  • Pk:
    50 g

Specifications

About this item

 :
 :195.08 g/mol
CAS Number:7440-06-4
Formula:Pt
MDL Number:MFCD00011179