Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Platinum
Platinum nominally 50% on carbon black
Platinum nominally 50% on carbon black
Platinum nominally 50% on carbon black
Catalog # 43989.14
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Expand image
Platinum nominally 50% on carbon black
Catalog # 43989.14
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Specifications
Pk:
25 g
Specifications
About this item
Product Details
Documentation
:
:
195.08 g/mol
CAS Number:
7440-06-4
Formula:
Pt
MDL Number:
MFCD00011179
Product Details
Documentation