AGI-6780 95%
Glass bottle, 100 mg
Catalog # ACRO467821000
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.
Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies
Synonyms: N-Cyclopropyl-4-(thiophen-3-yl)-3-(3-(3-(trifluoromethyl)phenyl)ureido)benzenesulphonamide
- CAS Number:1432660-47-3
- Formula:C₂₁H₁₈F₃N₃O₃S₂
- MDL Number:MFCD26097285
- Molecular Weight:481.51 g/mol
- Storage Temperature:Freezer
Specifications
Specification Test Results
- Appearance:White to yellow Powder or solid
- HPLC:≥94%
- MS:complies
- NMR (1H-NMR):Consistent with structure
Specifications
- Pack type:Glass bottle
- Pk:100 mg



