Order Entry
Export
ContactUsLinkComponent
Disinfectant spray, ClearKlens Tego® 2000
Disinfectant spray, ClearKlens Tego® 2000
Catalog # DIVY100961418
Supplier:  Diversey
Disinfectant spray, ClearKlens Tego® 2000
Catalog # DIVY100961418
Supplier:  Diversey

Specifications

  • Capacity:
    50×50 ml
  • Description:
    Disinfectant spray, Clearlens Tego® 2000, 50×50 ml
  • Environmentally Preferable:
  • Sterility:
    Sterile
  • Pk:
    50

Specifications

About this item

ClearKlens Tego® 2000 is a highly effective surface active amphoteric disinfectant with a broad spectrum of activity, developed for use in sterile environments.

  • Triple and double bagged
  • Sterile
  • 0.2 µm filtered
Certifications: CEN 1276, CEN 1650 and CEN 13697
Caution: Use biocides safely. Always read the label and product information before use.