Order Entry
Export
ContactUsLinkComponent
ADVASEP™-7, Sigma-Aldrich®
ADVASEP™-7, Sigma-Aldrich®
Catalog # A3723-250MG
CAS Number:  
ADVASEP™-7, Sigma-Aldrich®
Catalog # A3723-250MG
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Storage Temperature:Freezer