Mordant Blue 9 50% (dye content), Sigma-Aldrich®
Catalog # 195219-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:502.81 g/mol
Storage Temperature:Ambient
CAS Number:3624-68-8
Formula:C₁₆H₉ClN₂Na₂O₈S₂