Chrysophenine 65% (dye content), Sigma-Aldrich®
Catalog # 201901-50G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:680.66 g/mol
Storage Temperature:Ambient
CAS Number:2870-32-8
Formula:C₃₀H₂₈N₄O₈S₂
MDL Number:MFCD00007488