Acid Yellow 25 40% (dye content), Sigma-Aldrich®
Catalog # 201960-10G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:549.55 g/mol
Storage Temperature:Ambient
CAS Number:6359-85-9
Formula:C₂₃H₂N₅NaO₆S₂
MDL Number:MFCD00003121