Acid Red 18 (Food Red No. 102), Sigma-Aldrich®
Catalog # 199737-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:604.46 g/mol
Storage Temperature:Ambient
CAS Number:2611-82-7
Formula:C₂₀H₁₄N₂O₁₀S₃
MDL Number:MFCD00004084