
Astrazon Orange G, Sigma-Aldrich®
Catalog # 274216-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:350.88 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:3056-93-7
Formula:C₂₂H₂₃ClN₂
MDL Number:MFCD00011947