Order Entry
Export
ContactUsLinkComponent
N,N'-Bis(salicylidene)-1,2-phenylenediamine ≥97%, Sigma-Aldrich®
Irritant
N,N'-Bis(salicylidene)-1,2-phenylenediamine ≥97%, Sigma-Aldrich®
Catalog # 269492-5G
CAS Number:  
undefined
Irritant
N,N'-Bis(salicylidene)-1,2-phenylenediamine ≥97%, Sigma-Aldrich®
Catalog # 269492-5G
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :316.36 g/mol
Melting Pt:163…165 °C (lit.)
 :GHS07
Storage Temperature:Ambient
CAS Number:3946-91-6
Formula:C₂₀H₁₆N₂O₂
MDL Number:MFCD00009995