
Reactive black 5 ≥50 % (dye content), Sigma-Aldrich®
Catalog # 306452-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:991.789 g/mol
Melting Pt:>300 °C (lit.)
Density:1.21 g/cm³ (20 °C)
:GHS08
Storage Temperature:Ambient
CAS Number:17095-24-8
Formula:C₂₆H₂₁N₅Na₄O₁₉S₆