
N-(Diphenylmethylene)glycine ethyl ester 98%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:69555-14-2
:
Formula:C₁₇H₁₇NO₂
MDL Number:MFCD00010590
:267.32 g/mol
Melting Pt:51...53 °C (lit.)
:M-Clarity™ quality level = MQ200
Flash Pt:110 °C (closed cup)
:GHS08
Storage Temperature:Refrigerator