Order Entry
Export
ContactUsLinkComponent
Direct Red 23, Sigma-Aldrich®
Direct Red 23, Sigma-Aldrich®
Catalog # 212490-50G
CAS Number:  
undefined
Direct Red 23, Sigma-Aldrich®
Catalog # 212490-50G
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :813.72 g/mol
Storage Temperature:Ambient
CAS Number:3441-14-3
Formula:C₃₅H₂₅N₇Na₂OS₂
MDL Number:MFCD00003946