Cibacron Brilliant Yellow 3G-P, Sigma-Aldrich®
Catalog # 228478-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: Reactive yellow 2
:
:831.02 g/mol
Storage Temperature:Ambient
CAS Number:50662-99-2
Formula:C₂₅H₁₉Cl₃N₉NaOS₃
MDL Number:MFCD00009778