
Solvent Blue 59 98% (dye content), Sigma-Aldrich®
Catalog # 229121-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:294.35 g/mol
Melting Pt:215…217 °C (lit.)
:GHS07
Storage Temperature:Ambient
CAS Number:6994-46-3
Formula:C₁₈H₁₈N₂O₂
MDL Number:MFCD00071821