Order Entry
Export
ContactUsLinkComponent
Acid red 52, Sigma-Aldrich®
Acid red 52, Sigma-Aldrich®
Catalog # 230162-5G
CAS Number:  
undefined
Acid red 52, Sigma-Aldrich®
Catalog # 230162-5G
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :580.65 g/mol
Storage Temperature:Ambient
CAS Number:3520-42-1
Formula:C₂₇H₃₀N₂O₇S₂
MDL Number:MFCD00010180