Order Entry
Export
ContactUsLinkComponent
 
5-(Iodoacetamido)fluorescein ≥90% (by HPLC), Sigma-Aldrich®
 
undefined
5-(Iodoacetamido)fluorescein ≥90% (by HPLC), Sigma-Aldrich®

 

  • Appearance (Colour):
    Yellow to Orange
  • Appearance (Form):
    Powder
  • Carbon:
    48.5 - 54.1 %
  • Nitrogen:
    2.3 - 3.0 %
  • Purity (HPLC):
    ≥ 90 %
  • Solubility (Colour):
    Yellow to Orange
  • Solubility (Turbidity)
    5 mg/mL, DMSO:
    Clear