Phloxin B, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:18472-87-2
:
Formula:C₂₀H₂Br₄Cl₄Na₂O₅
MDL Number:MFCD00070627
:M-Clarity™ quality level = MQ200
:829.6344 g/mol
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Dark Red
- Appearance (Form):Powder
- Solubility (Colour):Red
- Solubility (Turbidity)
1 mg/mL, H2O:Clear - Infrared spectrum:Conforms to Structure
- E1%
In 50% Ethanol, at 548 nm:1150 - 1300 - Lambda Max:546 - 550 nm