
Direct Blue 15, Sigma-Aldrich®
Catalog # D2535-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:992.8 g/mol
:GHS08
Storage Temperature:Ambient
CAS Number:2429-74-5
Formula:C₃₄H₂₄N₆Na₄O₁₆S₄
MDL Number:MFCD00036441