Order Entry
Export
Orders LinkContactUsLinkComponent
 

Fast Garnet GBC Base solution, Sigma-Aldrich®

10 mL
Catalog # 3872-10ML
Health
Corrosive
Irritant
Fast Garnet GBC Base solution, Sigma-Aldrich®
Catalog # 3872-10ML
CAS Number:  
Fast Garnet GBC Base solution, Sigma-Aldrich®

10 mL

CAS Number:  
Price
203,00 €
Log in for availability
1 * 10 mL
Product Details & Documents

Intended for use in Sigma Procedure 387 for the demonstration of Acid Phosphatase (TRAP).

  • MDL Number:MFCD00007733
  • Storage Temperature:Refrigerator
  • ADR:8,II
Specifications

Specifications

  • Pk:
    10 mL

Specifications