Order Entry
Export
ContactUsLinkComponent
Zinc 10% (w/v) in THF, suspension
Flammable
Irritant
Health
Environment
Zinc 10% (w/v) in THF, suspension
Catalog # ACRO463731000
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
Zinc 10% (w/v) in THF, suspension
Catalog # ACRO463731000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Suspension
  • Strength:
    9.8 to 10.2 % (w/v)

Specification Test Results

About this item

 :
 :65.39 g/mol
Density:0.949
Flash Pt:–17 °C
Storage Temperature:Refrigerator
UN:3399
ADR:4.3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291