Letrozole
Glass bottle, 1 g
Catalog # ACRO463190010
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- Bioactive
Synonyms: 1-Bis(4-cyanophenyl)methyl-1,2,4-triazole, 4,4'-(1,2,4-Triazol-1-ylmethylene)dibenzonitrile
- CAS Number:112809-51-5
- Formula:C₁₇H₁₁N₅
- MDL Number:MFCD00866241
- Molecular Weight:285.31 g/mol
- Melting Pt:185 °C
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:183 to 187 °C
- Water:≤0.5 %
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



