
1,2-Bis(trichlorosilyl)ethane
:2504-64-5
:
:C₂H₄Cl₆Si₂
:MFCD00013601
:296.92 g/mol
:24…25 °C
:65 °C
:Ambient
:2987
:8,II
:GHS05
- Appearance:Colorless to light yellow Crystalline mass or liquid
- Infrared spectrum:Conforms
- Refractive index:1.4735 to 1.4760 (20°C, 589 nm)
- GC:≥94 %