Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Carbolic fuchsin
Carbolic fuchsin Ziehl 1/10
Carbolic fuchsin Ziehl 1/10
Carbolic fuchsin Ziehl 1/10
Catalog # RALA364540-1000
Supplier:
RAL
CAS Number:
Expand image
Carbolic fuchsin Ziehl 1/10
Catalog # RALA364540-1000
Supplier:
RAL
CAS Number:
Specifications
Pk:
6
Seq:
0006
Specifications
About this item
Product Details
:
:
1
Product Details