Order Entry
Export
Orders LinkContactUsLinkComponent
FLUO-RAL Kit Fluorescent Staining

FLUO-RAL Kit Fluorescent Staining

1 kit, 1
Catalog # RALA359000-0000
Supplier:   RAL
FLUO-RAL Kit Fluorescent Staining
Catalog # RALA359000-0000
Supplier:  RAL
CAS Number:  
FLUO-RAL Kit Fluorescent Staining

1 kit, 1

CAS Number:  
Price
107,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Kit Fluo-RAL offers a rapid method for the identification of mycobacteria or Acid Fast Bacilli (AFB). The penetration by the discolouring agent of the highly specific structure of the wall is hardly possible.

This property enables the AFB to retain the fluorescent yellow-green colour of the auramine after discolouration with acid and alcohol. The other non-acid-alcohol-resistant bacteria and cell elements are counter-stained red thanks to Thiazine Red.

Components - Name (ref) Packing
FLUO-RAL FIXATIVE (ref 362115) 250 ml (Other packaging 1000 ml)
AURAMIN FLUO-RAL (ref 362325) 125 ml (Other packaging 500 ml)
CARBOLIC SOLUTION FOR FLUO-RAL AURAMIN (ref 362530) 125 ml (Other packaging 500 ml)
FLUO-RAL DEGOMMIER DISCOLOURING (Ref 362210) 250 ml (Other packaging 1000 ml)
FLUO-RAL THIAZIN RED (Ref 362780) 125 ml (Other packaging 500 ml)
CARBOLIC SOLUTION FOR FLUO-RAL THIAZIN RED (Ref 362610) 125 ml (Other packaging 500 ml)

Synonyms:  FAI, Formamidinium iodide

Documents

Specifications

Specifications

  • Pk:
    1
  • Pk Info:
    1 kit

Specifications